Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cardiac mesoderm enhancer-associated non-coding RNA | CARMN |
CARMEN, MIR143HG |
-66.2kb | Anti-sense | Chr. 5 - 149406845 - 149432836 on NCBI Build GRCh38 |
| Downstream | Rho guanine nucleotide exchange factor 37 | ARHGEF37 | 52.2kb | Anti-sense | Chr. 5 - 149552049 - 149634968 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map