Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105371708 | LOC105371708 | -142.0kb | Sense | Chr. 17 - 27928924 - 27991731 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC105371930 | LOC105371930 | 88.7kb | Sense | Chr. 17 - 28222783 - 28228461 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map