Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | pyruvate dehydrogenase (lipoamide) alpha 2 | PDHA2 | PDHAL | -44.1kb | Anti-sense | Chr. 4 - 95840088 - 95841474 on NCBI Build GRCh38 |
| Downstream | STPG2 antisense RNA 1 | STPG2-AS1 | 1.5mb | Anti-sense | Chr. 4 - 97366926 - 97490164 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map