Assay Details
| Gene Symbol: RNA18S5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003286.2 | 1 | 1257 | 152 | ||
| Gene Symbol: RNA45S5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046235.1 | 1 | 4911 | 152 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3390107 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4161 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4162 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 59328 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3390107 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3390107 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_003286_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7954311 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7954310 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC22_KI270733v1_random00006435.hg.1 | ENST00000610594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC22_KI270733v1_random00006435.hg.1 | ENST00000611782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC22_KI270733v1_random00006435.hg.1 | ENST00000611802.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC22_KI270733v1_random00006435.hg.1 | ENST00000611919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC22_KI270733v1_random00006435.hg.1 | ENST00000611934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC22_KI270733v1_random00006435.hg.1 | ENST00000616292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006476.hg.1 | ENST00000610460.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006597.hg.1 | ENST00000515896.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008794.hg.1 | ENST00000614365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010228.hg.1 | ENST00000365096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TCUn_KI270442v100006434.hg.1 | ENST00000611690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006488.hg.1 | ENST00000612173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006488.hg.1 | ENST00000613635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006488.hg.1 | ENST00000614525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006488.hg.1 | ENST00000616418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006488.hg.1 | ENST00000616457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006488.hg.1 | ENST00000619471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006485.hg.1 | ENST00000613359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007033.hg.1 | ENST00000445125.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007033.hg.1 | ENST00000445125.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007033.hg.1 | ENST00000445125.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007033.hg.1 | ENST00000445125.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007033.hg.1 | ENST00000445125.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007033.hg.1 | ENST00000445125.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007033.hg.1 | ENST00000445125.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007650.hg.1 | ENST00000363564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TCUn_GL000220v100006432.hg.1 | ENST00000611431.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TCUn_GL000220v100006432.hg.1 | ENST00000611446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TCUn_GL000220v100006432.hg.1 | ENST00000614470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TCUn_GL000220v100006432.hg.1 | ENST00000614928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TCUn_GL000220v100006432.hg.1 | ENST00000615115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TCUn_GL000220v100006432.hg.1 | ENST00000617883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TCUn_GL000220v100006432.hg.1 | ENST00000619779.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC22_KI270733v1_random00006442.hg.1 | ENST00000618998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007833.hg.1 | ENST00000612732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006469.hg.1 | ENST00000612463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006469.hg.1 | NR_003285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC22_KI270733v1_random00006442.hg.1 | ENST00000618998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TCUn_KI270442v100006434.hg.1 | ENST00000611690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100144325.hg.1 | ENST00000619471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100144325.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100144325.hg.1 | TC2100006488.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100144325.hg.1 | ENST00000619471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100144325.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006469.hg.1 | ENST00000612463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006469.hg.1 | NR_003285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006488.hg.1 | ENST00000612173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006488.hg.1 | ENST00000613635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006488.hg.1 | ENST00000614525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006488.hg.1 | ENST00000616418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006488.hg.1 | ENST00000616457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006488.hg.1 | ENST00000619471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006488.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC22_KI270733v1_random00006435.hg.1 | ENST00000610594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC22_KI270733v1_random00006435.hg.1 | ENST00000611782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC22_KI270733v1_random00006435.hg.1 | ENST00000611802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC22_KI270733v1_random00006435.hg.1 | ENST00000611919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC22_KI270733v1_random00006435.hg.1 | ENST00000611934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC22_KI270733v1_random00006435.hg.1 | ENST00000616292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100144327.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100144327.hg.1 | TC2100006488.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100144327.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007833.hg.1 | ENST00000612732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010228.hg.1 | ENST00000365096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006476.hg.1 | ENST00000610460.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007033.hg.1 | ENST00000445125.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007033.hg.1 | ENST00000445125.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007033.hg.1 | ENST00000445125.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007033.hg.1 | ENST00000445125.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007033.hg.1 | ENST00000445125.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007033.hg.1 | ENST00000445125.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007033.hg.1 | ENST00000445125.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007650.hg.1 | ENST00000363564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TCUn_GL000220v100006432.hg.1 | ENST00000611431.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TCUn_GL000220v100006432.hg.1 | ENST00000611446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TCUn_GL000220v100006432.hg.1 | ENST00000614470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TCUn_GL000220v100006432.hg.1 | ENST00000614928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TCUn_GL000220v100006432.hg.1 | ENST00000615115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TCUn_GL000220v100006432.hg.1 | ENST00000617883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TCUn_GL000220v100006432.hg.1 | ENST00000619779.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100144324.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100144324.hg.1 | TC2100006488.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100144324.hg.1 | NR_046235.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00006597.hg.1 | ENST00000515896.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008794.hg.1 | ENST00000614365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006485.hg.1 | ENST00000613359.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 148950 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 187876 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3243593 | X03205.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3243593 | NR_003286.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3243593 | AK027322.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3243593 | X03205.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3243593 | NR_003286.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3243593 | AK027322.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3243593 | X03205.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3243593 | NR_003286.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3243593 | AK027322.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3243593 | X03205.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3243593 | NR_003286.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3243593 | AK027322.1 | human_transcriptome |

RNA18S5
Genomic Map