Assay Details
| Gene Symbol: NODAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018055.4 | NP_060525.3 | 2 | 783 | 64 | |
| GenBank mRNA | BC033585.1 | - | 2 | 677 | 64 | |
| BC104976.1 | - | 2 | 783 | 8143979 | 64 | |
| BC112025.1 | - | 2 | 795 | 8327493 | 64 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NODAL
Genomic Map