| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000510661.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000510661.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000510661.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000510661.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000510661.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000631833.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000631833.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000631833.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000631833.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000632230.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000632230.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000632230.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005378.hg.1 |
TC0X000393.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005378.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005378.hg.1 |
ENST00000510661.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005378.hg.1 |
ENST00000535692.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005378.hg.1 |
ENST00000540800.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005378.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005380.hg.1 |
TC0X000393.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005380.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005380.hg.1 |
ENST00000510661.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005380.hg.1 |
ENST00000535692.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005380.hg.1 |
ENST00000540800.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005380.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00177666.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00177666.hg.1 |
ENST00000510661.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00177666.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00177666.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00177666.hg.1 |
ENST00000510661.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00177666.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00177666.hg.1 |
TC0X00011305.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00177664.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00177664.hg.1 |
ENST00000510661.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00177664.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00177664.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00177664.hg.1 |
ENST00000510661.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00177664.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00177664.hg.1 |
TC0X00011305.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000510661.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000510661.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000510661.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000510661.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000510661.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000631833.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000631833.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000631833.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000631833.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000632230.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000632230.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000632230.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |