Assay Details
| Gene Symbol: ITM2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171581.1 | NP_001165052.1 | 2 - 3 | 646 | 82 | |
| NM_004867.4 | NP_004858.1 | 3 - 4 | 778 | 82 | ||
| GenBank mRNA | AF038953.1 | - | 3 - 4 | 580 | 82 | |
| AK223327.1 | - | 3 - 4 | 557 | 82 | ||
| AK303238.1 | - | 2 - 3 | 428 | 82 | ||
| AK313154.1 | - | 3 - 4 | 736 | 82 | ||
| AY359051.1 | - | 3 - 4 | 519 | 82 | ||
| BC040437.1 | - | 3 - 4 | 533 | 5748519 | 82 | |
| CR456875.1 | - | 3 - 4 | 441 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001180.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015894.hg.1 | ENST00000373298.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015894.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015894.hg.1 | ENST00000469541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015894.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015894.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015894.hg.1 | TC0X001180.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000461357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000461357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000461357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000462038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000462038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000462038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000461357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000461357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000461357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000462038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000462038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000462038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00169276.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169276.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169276.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169276.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169276.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00169276.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00169276.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169276.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169276.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169276.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169276.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00169276.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00169276.hg.1 | TC0X00010155.hg.1 | Clariom D TC |

ITM2A
Genomic Map