Assay Details
| Gene Symbol: PALM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037293.2 | NP_001032370.1 | 7 | 1148 | 107 | |
| NM_053016.5 | NP_443749.5 | 7 | 1362 | 107 | ||
| GenBank mRNA | AJ312216.1 | - | 7 | 1220 | 107 | |
| AK095003.1 | - | 7 | 1114 | 107 | ||
| BC039306.1 | - | 7 | 1150 | 5260371 | 107 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000314527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000314527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000314527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000314527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000314527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000314527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000314527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000314527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000314527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000314527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000374531.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000374531.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000374531.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000374531.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000374531.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000374531.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000374531.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000374531.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000374531.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000374531.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000374531.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000465091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000465091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000465091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000465091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000483909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000483909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000483909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000483909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000483909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000483909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000483909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000483909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000483909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000483909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000483909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000497711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000497711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000497711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000497711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000497711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | ENST00000497711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000314527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000314527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000314527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000314527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000314527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000314527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000314527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000314527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000314527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000314527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000374531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000374531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000374531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000374531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000374531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000374531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000374531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000374531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000374531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000374531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000374531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000465091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000465091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000465091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000465091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000483909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000483909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000483909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000483909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000483909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000483909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000483909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000483909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000483909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000483909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000483909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000497711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000497711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000497711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000497711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000497711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | ENST00000497711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_001037293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_053016.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012156.hg.1 | NM_053016.5 | Refseq |

PALM2
Genomic Map