Assay Details
| Gene Symbol: NUDT16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171906.1 | NP_001165377.1 | 2 | 650 | 101 | |
| GenBank mRNA | AK304650.1 | - | 2 | 621 | 101 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000710.hg.1
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
adenosine to inosine editing
mRNA catabolic process
proteolysis
positive regulation of cell proliferation
snoRNA catabolic process
dephosphorylation
nucleobase-containing small molecule catabolic process
dITP catabolic process
IDP catabolic process
positive regulation of cell cycle process
XDP catabolic process
negative regulation of rRNA processing
positive regulation of double-strand break repair
magnesium ion binding
mRNA binding
GTP binding
metalloexopeptidase activity
manganese ion binding
snoRNA binding
dITP diphosphatase activity
identical protein binding
protein homodimerization activity
m7G(5')pppN diphosphatase activity
cobalt ion binding
inosine-diphosphatase activity
dIDP diphosphatase activity
nucleotide phosphatase activity, acting on free nucleotides
XTP binding
ITP binding
adenosine to inosine editing
mRNA catabolic process
proteolysis
positive regulation of cell proliferation
snoRNA catabolic process
dephosphorylation
nucleobase-containing small molecule catabolic process
dITP catabolic process
IDP catabolic process
positive regulation of cell cycle process
XDP catabolic process
negative regulation of rRNA processing
positive regulation of double-strand break repair
magnesium ion binding
mRNA binding
GTP binding
metalloexopeptidase activity
manganese ion binding
snoRNA binding
dITP diphosphatase activity
identical protein binding
protein homodimerization activity
m7G(5')pppN diphosphatase activity
cobalt ion binding
inosine-diphosphatase activity
dIDP diphosphatase activity
nucleotide phosphatase activity, acting on free nucleotides
XTP binding
ITP binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03012760.hg.1 | ENST00000502852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03012760.hg.1 | NM_001171906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03012760.hg.1 | TC03000710.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | ENST00000502852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | ENST00000502852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | ENST00000502852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | ENST00000502852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | ENST00000502852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | ENST00000502852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | ENST00000502852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | ENST00000502852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | ENST00000502852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | ENST00000521288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | ENST00000521288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | ENST00000521288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | ENST00000521288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | ENST00000521288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | ENST00000521288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | ENST00000521288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | ENST00000537561.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | ENST00000537561.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | ENST00000537561.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | ENST00000537561.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | ENST00000537561.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | ENST00000537561.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NM_001171905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NM_001171905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NM_001171905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NM_001171905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NM_001171905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NM_001171905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NM_001171906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NM_001171906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NM_001171906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NM_001171906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NM_152395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NM_152395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NM_152395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NM_152395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NM_152395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NM_152395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NM_152395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NR_033268.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NR_033268.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NR_033268.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NR_033268.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NR_033268.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NR_033268.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NR_033268.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008830.hg.1 | NR_033268.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300163002.hg.1 | ENST00000502852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163002.hg.1 | NM_001171906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300163002.hg.1 | ENST00000502852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163002.hg.1 | NM_001171906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300163002.hg.1 | TC0300008830.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | ENST00000502852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | ENST00000502852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | ENST00000502852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | ENST00000502852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | ENST00000502852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | ENST00000502852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | ENST00000502852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | ENST00000502852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | ENST00000502852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | ENST00000521288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | ENST00000521288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | ENST00000521288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | ENST00000521288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | ENST00000521288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | ENST00000521288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | ENST00000521288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | ENST00000537561.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | ENST00000537561.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | ENST00000537561.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | ENST00000537561.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | ENST00000537561.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | ENST00000537561.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NM_001171905.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NM_001171905.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NM_001171905.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NM_001171905.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NM_001171905.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NM_001171905.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NM_001171906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NM_001171906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NM_001171906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NM_001171906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NM_152395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NM_152395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NM_152395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NM_152395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NM_152395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NM_152395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NM_152395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NR_033268.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NR_033268.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NR_033268.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NR_033268.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NR_033268.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NR_033268.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NR_033268.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008830.hg.1 | NR_033268.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300163003.hg.1 | ENST00000502852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163003.hg.1 | NM_001171906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300163003.hg.1 | ENST00000502852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163003.hg.1 | NM_001171906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300163003.hg.1 | TC0300008830.hg.1 | Clariom D TC |

NUDT16
Genomic Map