Assay Details
| Gene Symbol: EPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000799.2 | NP_000790.2 | 3 - 4 | 431 | 134 | |
| GenBank mRNA | BC093628.1 | - | 3 - 4 | 328 | 7939473 | 134 |
| BC111937.1 | - | 3 - 4 | 328 | 8327405 | 134 | |
| BC143225.1 | - | 3 - 4 | 328 | 9051726 | 134 | |
| JN849371.1 | - | 3 - 4 | 250 | 134 | ||
| KF178447.1 | - | 2 - 3 | 259 | 134 | ||
| S65458.1 | - | 1 - 2 | 81 | 134 | ||
| X02157.1 | - | 3 - 4 | 431 | 134 | ||
More Information
Set Membership: |
|

EPO
Genomic Map