Assay Details
| Gene Symbol: MRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002438.3 | NP_002429.1 | 14 - 15 | 2321 | 108 | |
| GenBank mRNA | AK308167.1 | - | 14 - 15 | 2317 | 108 | |
| DQ663787.1 | - | 14 - 15 | 2198 | 108 | ||
| J05550.1 | - | 14 - 15 | 2301 | 108 | ||
| X55635.1 | - | 14 - 15 | 2274 | 108 | ||
More Information
Set Membership: |
|

MRC1
Genomic Map