| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001188.hg.1 |
ENST00000334534.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001188.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001188.hg.1 |
TC20000083.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001191.hg.1 |
ENST00000334534.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001191.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001191.hg.1 |
TC20000083.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001190.hg.1 |
ENST00000334534.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001190.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001190.hg.1 |
TC20000083.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006694.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006694.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006694.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006694.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006694.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006694.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006694.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006694.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006694.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006694.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006694.hg.1 |
ENST00000488816.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006694.hg.1 |
ENST00000488816.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006694.hg.1 |
ENST00000488816.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006694.hg.1 |
ENST00000488816.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006694.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006694.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006694.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006694.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006694.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006694.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006694.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006694.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006694.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001187.hg.1 |
ENST00000334534.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001187.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001187.hg.1 |
TC20000083.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001189.hg.1 |
ENST00000334534.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001189.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001189.hg.1 |
TC20000083.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2000145909.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145909.hg.1 |
ENST00000488816.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145909.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000145909.hg.1 |
TC2000006694.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000145909.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145909.hg.1 |
ENST00000488816.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145909.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000145902.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145902.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000145902.hg.1 |
TC2000006694.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000145902.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145902.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000145903.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145903.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000145903.hg.1 |
TC2000006694.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000145903.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145903.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000145908.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145908.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000145908.hg.1 |
TC2000006694.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000145908.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145908.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000145904.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145904.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000145904.hg.1 |
TC2000006694.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000145904.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145904.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000145906.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145906.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000145906.hg.1 |
TC2000006694.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000145906.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145906.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006694.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006694.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006694.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006694.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006694.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006694.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006694.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006694.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006694.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006694.hg.1 |
ENST00000334534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006694.hg.1 |
ENST00000488816.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006694.hg.1 |
ENST00000488816.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006694.hg.1 |
ENST00000488816.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006694.hg.1 |
ENST00000488816.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006694.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006694.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006694.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006694.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006694.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006694.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006694.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006694.hg.1 |
NM_001009608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006694.hg.1 |
NM_001009608.2 |
Refseq |