Assay Details
| Gene Symbol: SLX4IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009608.2 | NP_001009608.1 | 7 - 8 | 690 | 69 | |
| XM_017027647.1 | XP_016883136.1 | 6 - 7 | 4192 | 69 | ||
| XM_017027648.1 | XP_016883137.1 | 8 - 9 | 1643 | 69 | ||
| XM_017027649.1 | XP_016883138.1 | 8 - 9 | 836 | 69 | ||
| XM_017027650.1 | XP_016883139.1 | 8 - 9 | 1603 | 69 | ||
| XM_017027651.1 | XP_016883140.1 | 5 - 6 | 641 | 69 | ||
| XM_017027652.1 | XP_016883141.1 | 7 - 8 | 1592 | 69 | ||
| GenBank mRNA | BC020787.1 | - | 7 - 8 | 690 | 69 | |
| BC026094.1 | - | 7 - 8 | 694 | 69 | ||
More Information
Set Membership: |
|

SLX4IP
Genomic Map