Assay Details
| Gene Symbol: AADACL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103170.2 | NP_001096640.2 | 3 - 4 | 513 | 65 | |
| NR_111984.1 | 2 - 3 | 296 | 65 | |||
| GenBank mRNA | AK127828.1 | - | 2 - 3 | 296 | 65 | |
| BG697255.1 | - | 3 - 4 | 485 | 65 | ||
More Information
Set Membership: |
|

AADACL3
Genomic Map