Assay Details
| Gene Symbol: ZBED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171136.1 | NP_001164607.1 | 1 - 2 | 150 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001171136.1 transcripts.
Set Membership: |
|

ZBED1
Genomic Map