Assay Details
| Gene Symbol: ZNF300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172832.1 | NP_001166303.1 | 2 - 3 | 393 | 65 | |
| GenBank mRNA | DB094933.1 | - | 2 - 3 | 386 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001172832.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_052860.2 | Refseq |

ZNF300
Genomic Map