| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19012827.hg.1 |
ENST00000360338.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19012827.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19012827.hg.1 |
TC19000937.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19012826.hg.1 |
ENST00000360338.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19012826.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19012826.hg.1 |
TC19000937.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008989.hg.1 |
ENST00000360338.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008989.hg.1 |
ENST00000360338.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008989.hg.1 |
ENST00000360338.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008989.hg.1 |
ENST00000360338.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008989.hg.1 |
ENST00000360338.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008989.hg.1 |
ENST00000360338.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008989.hg.1 |
ENST00000360338.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008989.hg.1 |
ENST00000537645.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008989.hg.1 |
ENST00000537645.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008989.hg.1 |
ENST00000537645.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008989.hg.1 |
ENST00000537645.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008989.hg.1 |
ENST00000537645.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008989.hg.1 |
ENST00000599602.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008989.hg.1 |
ENST00000599602.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008989.hg.1 |
ENST00000599602.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008989.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008989.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008989.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008989.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008989.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008989.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008989.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008989.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19012828.hg.1 |
ENST00000360338.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19012828.hg.1 |
ENST00000537645.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19012828.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19012828.hg.1 |
TC19000937.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900170734.hg.1 |
ENST00000360338.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900170734.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900170734.hg.1 |
TC1900008989.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900170734.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900170734.hg.1 |
ENST00000360338.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008989.hg.1 |
ENST00000360338.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008989.hg.1 |
ENST00000360338.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008989.hg.1 |
ENST00000360338.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008989.hg.1 |
ENST00000360338.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008989.hg.1 |
ENST00000360338.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008989.hg.1 |
ENST00000360338.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008989.hg.1 |
ENST00000360338.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008989.hg.1 |
ENST00000537645.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008989.hg.1 |
ENST00000537645.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008989.hg.1 |
ENST00000537645.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008989.hg.1 |
ENST00000537645.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008989.hg.1 |
ENST00000537645.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008989.hg.1 |
ENST00000599602.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008989.hg.1 |
ENST00000599602.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008989.hg.1 |
ENST00000599602.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008989.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008989.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008989.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008989.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008989.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008989.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008989.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008989.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900170735.hg.1 |
ENST00000360338.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900170735.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900170735.hg.1 |
TC1900008989.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900170735.hg.1 |
ENST00000360338.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900170735.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900170737.hg.1 |
ENST00000360338.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900170737.hg.1 |
ENST00000537645.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900170737.hg.1 |
ENST00000599602.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900170737.hg.1 |
NM_006635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900170737.hg.1 |
TC1900008989.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900170737.hg.1 |
ENST00000360338.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900170737.hg.1 |
ENST00000537645.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900170737.hg.1 |
ENST00000599602.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900170737.hg.1 |
NM_006635.3 |
Refseq |