Assay Details
| Gene Symbol: ZNF460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006635.3 | NP_006626.3 | 2 - 3 | 477 | 77 | |
| XM_005258422.3 | XP_005258479.1 | 2 - 3 | 842 | 77 | ||
| XM_017026176.1 | XP_016881665.1 | 2 - 3 | 292 | 77 | ||
| GenBank mRNA | AK074582.1 | - | 2 - 3 | 472 | 77 | |
| AK292144.1 | - | 2 - 3 | 477 | 77 | ||
| AK298106.1 | - | 2 - 3 | 199 | 77 | ||
| AY329492.1 | - | 2 - 3 | 398 | 77 | ||
| BC109018.1 | - | 2 - 3 | 155 | 77 | ||
| BC109019.1 | - | 2 - 3 | 155 | 77 | ||
| BC118625.1 | - | 2 - 3 | 155 | 40006201 | 77 | |
More Information
Set Membership: |
|

ZNF460
Genomic Map