Assay Details
| Gene Symbol: TRIL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014817.3 | NP_055632.2 | 1 | 2557 | 71 | |
| GenBank mRNA | AB014544.1 | - | 1 | 2436 | 71 | |
| BC036337.1 | - | 1 | 2462 | 5179761 | 71 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001235.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07019279.hg.1 | ENST00000322982.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07019279.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07019279.hg.1 | TC07001235.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07019277.hg.1 | ENST00000322982.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07019277.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07019277.hg.1 | TC07001235.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010603.hg.1 | ENST00000322982.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010603.hg.1 | ENST00000322982.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010603.hg.1 | ENST00000322982.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010603.hg.1 | ENST00000322982.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010603.hg.1 | ENST00000322982.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010603.hg.1 | ENST00000539664.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010603.hg.1 | ENST00000539664.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010603.hg.1 | ENST00000539664.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010603.hg.1 | ENST00000539664.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010603.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010603.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010603.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010603.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010603.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010603.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000436594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000436594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000436594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000436594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000608972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000608972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000608972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000608972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | ENST00000609495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | NR_038965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | NR_038965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | NR_038965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | NR_038965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | NR_038965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | NR_038965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | NR_038965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | NR_038965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007038.hg.1 | NR_038965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07019282.hg.1 | ENST00000322982.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07019282.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07019282.hg.1 | TC07001235.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07019283.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07019283.hg.1 | TC07001235.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0700175359.hg.1 | ENST00000322982.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175359.hg.1 | ENST00000539664.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175359.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700175359.hg.1 | ENST00000322982.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175359.hg.1 | ENST00000539664.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175359.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700175359.hg.1 | TC0700010603.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700175361.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700175361.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700175361.hg.1 | TC0700010603.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700175358.hg.1 | ENST00000322982.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175358.hg.1 | ENST00000539664.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175358.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700175358.hg.1 | ENST00000322982.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175358.hg.1 | ENST00000539664.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175358.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700175358.hg.1 | TC0700010603.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700175360.hg.1 | ENST00000322982.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175360.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700175360.hg.1 | ENST00000322982.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175360.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700175360.hg.1 | TC0700010603.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000436594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000436594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000436594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000436594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000608972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000608972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000608972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000608972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | ENST00000609495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | NR_038965.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | NR_038965.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | NR_038965.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | NR_038965.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | NR_038965.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | NR_038965.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | NR_038965.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | NR_038965.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007038.hg.1 | NR_038965.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700175357.hg.1 | ENST00000322982.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175357.hg.1 | ENST00000539664.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175357.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700175357.hg.1 | ENST00000322982.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175357.hg.1 | ENST00000539664.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175357.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700175357.hg.1 | TC0700010603.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700010603.hg.1 | ENST00000322982.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010603.hg.1 | ENST00000322982.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010603.hg.1 | ENST00000322982.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010603.hg.1 | ENST00000322982.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010603.hg.1 | ENST00000322982.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010603.hg.1 | ENST00000539664.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010603.hg.1 | ENST00000539664.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010603.hg.1 | ENST00000539664.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010603.hg.1 | ENST00000539664.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010603.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010603.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010603.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010603.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010603.hg.1 | NM_014817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010603.hg.1 | NM_014817.3 | Refseq |

TRIL
Genomic Map