Assay Details
| Gene Symbol: GRAPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129778.1 | NP_001123250.1 | 3 - 4 | 443 | 109 | |
| XM_017024637.1 | XP_016880126.1 | 4 - 5 | 522 | 109 | ||
| GenBank mRNA | BC157869.1 | - | 3 - 4 | 443 | 9057088 | 109 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000229.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P299355 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 44655 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 43128 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P392962 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P299355 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P299355 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P299355 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8005551 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8005556 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8013328 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8005550 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8005552 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8013327 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8005549 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8005555 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1560524_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1559689_PM_a_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 239074_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1559688_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11738004_s_at | - | - |
| Affymetrix Human Genome U219 Array | 11738004_PM_s_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000437957.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000437957.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000437957.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000437957.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000574324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000574324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000574324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000574324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000574324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000574324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000577213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000577213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000577213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000577213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000577213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000577213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000583540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000583540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000583540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | ENST00000583540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007153.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003274.hg.1 | ENST00000344415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003274.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003274.hg.1 | TC17000229.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700150756.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150756.hg.1 | ENST00000574324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150756.hg.1 | ENST00000583540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150756.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700150756.hg.1 | TC1700007153.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700150756.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700150756.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150756.hg.1 | ENST00000574324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150756.hg.1 | ENST00000583540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150757.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150757.hg.1 | ENST00000574324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150757.hg.1 | ENST00000583540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150757.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700150757.hg.1 | TC1700007153.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700150757.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150757.hg.1 | ENST00000574324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150757.hg.1 | ENST00000583540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150757.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000437957.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000437957.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000437957.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000437957.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000574324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000574324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000574324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000574324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000574324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000574324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000577213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000577213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000577213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000577213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000577213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000577213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000583540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000583540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000583540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | ENST00000583540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007153.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700150758.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150758.hg.1 | ENST00000574324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150758.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700150758.hg.1 | TC1700007153.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700150758.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150758.hg.1 | ENST00000574324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150758.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700150752.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150752.hg.1 | ENST00000437957.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150752.hg.1 | ENST00000574324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150752.hg.1 | ENST00000577213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150752.hg.1 | ENST00000583540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150752.hg.1 | NM_001129778.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700150752.hg.1 | TC1700007153.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700150752.hg.1 | ENST00000344415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150752.hg.1 | ENST00000437957.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150752.hg.1 | ENST00000574324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150752.hg.1 | ENST00000577213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150752.hg.1 | ENST00000583540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150752.hg.1 | NM_001129778.2 | Refseq |
| Human Genome U133 Plus 2.0 Array | 239074_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1560524_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1559688_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1559688_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1560524_at | - | - |
| Human Genome U133 Plus 2.0 Array | 244666_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1559689_PM_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1559689_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 239074_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 244666_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 239074_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 239074_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559689_a_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560524_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559688_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 239074_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559689_a_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559689_a_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559688_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560524_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 239074_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 239074_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 239074_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559689_a_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559688_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559688_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560524_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560524_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560524_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559689_a_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 239074_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559688_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559689_a_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559688_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 239074_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559688_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559688_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559688_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 239074_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559689_a_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559688_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560524_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 239074_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559689_a_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560524_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559689_a_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560524_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559689_a_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560524_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560524_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559689_a_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560524_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 239074_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559688_at11 | - | - |
| Human Genome U95 Set | 48298_PM_at | - | - |
| Human Genome U95 Set | 48298_at | - | - |
| Human Genome U133 Set | 239074_at | - | - |
| Human Genome U133 Set | 239074_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009224 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009224 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 132075 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1729680 | BC026233.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1729680 | BC060883.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1729680 | AK026312.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1729680 | BC026233.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1729680 | BC060883.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1729680 | AK026312.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1656472 | BC157869.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1656472 | NM_001129778.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1656472 | CR595033.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1656472 | BQ056077.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1656472 | NM_006613.3 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1656472 | BC157869.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1656472 | NM_001129778.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1656472 | CR595033.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1656472 | BQ056077.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1656472 | NM_006613.3 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1729680 | BC026233.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1729680 | BC060883.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1729680 | AK026312.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1729680 | BC026233.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1729680 | BC060883.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1729680 | AK026312.1 | human_transcriptome |

GRAPL
Genomic Map