Assay Details
| Gene Symbol: SETBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130110.1 | NP_001123582.1 | 1 - 2 | 222 | 94 | |
| GenBank mRNA | BC062338.1 | - | 1 - 2 | 94 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000160.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC062338.1,NM_001130110.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR18001855.hg.1 | ENST00000426838.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001855.hg.1 | NM_001130110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001855.hg.1 | TC18000160.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001858.hg.1 | ENST00000282030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001858.hg.1 | ENST00000426838.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001858.hg.1 | NM_001130110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001858.hg.1 | NM_015559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001858.hg.1 | TC18000160.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | ENST00000282030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | ENST00000282030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | ENST00000282030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | ENST00000282030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | ENST00000282030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | ENST00000282030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | ENST00000282030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | ENST00000282030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | ENST00000282030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | ENST00000282030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | ENST00000426838.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | ENST00000426838.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | ENST00000426838.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | ENST00000426838.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | ENST00000426838.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | ENST00000426838.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | ENST00000426838.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | ENST00000591940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | ENST00000591940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | ENST00000591940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | NM_001130110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | NM_001130110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | NM_001130110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | NM_001130110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | NM_001130110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | NM_001130110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | NM_001130110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | NM_015559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | NM_015559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | NM_015559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | NM_015559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | NM_015559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | NM_015559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | NM_015559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | NM_015559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | NM_015559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007186.hg.1 | NM_015559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | ENST00000282030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | ENST00000282030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | ENST00000282030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | ENST00000282030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | ENST00000282030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | ENST00000282030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | ENST00000282030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | ENST00000282030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | ENST00000282030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | ENST00000282030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | ENST00000426838.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | ENST00000426838.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | ENST00000426838.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | ENST00000426838.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | ENST00000426838.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | ENST00000426838.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | ENST00000426838.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | ENST00000591940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | ENST00000591940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | ENST00000591940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | NM_001130110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | NM_001130110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | NM_001130110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | NM_001130110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | NM_001130110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | NM_001130110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | NM_001130110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | NM_015559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | NM_015559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | NM_015559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | NM_015559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | NM_015559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | NM_015559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | NM_015559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | NM_015559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | NM_015559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007186.hg.1 | NM_015559.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800149291.hg.1 | ENST00000282030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800149291.hg.1 | ENST00000426838.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800149291.hg.1 | ENST00000591940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800149291.hg.1 | NM_001130110.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800149291.hg.1 | NM_015559.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800149291.hg.1 | TC1800007186.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800149291.hg.1 | ENST00000282030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800149291.hg.1 | ENST00000426838.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800149291.hg.1 | ENST00000591940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800149291.hg.1 | NM_001130110.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800149291.hg.1 | NM_015559.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800149287.hg.1 | ENST00000426838.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800149287.hg.1 | NM_001130110.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800149287.hg.1 | TC1800007186.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800149287.hg.1 | ENST00000426838.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800149287.hg.1 | NM_001130110.1 | Refseq |

SETBP1
Genomic Map