Assay Details
| Gene Symbol: AKAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136562.2 | NP_001130034.1 | 1 - 2 | 96 | 101 | |
| GenBank mRNA | AK300427.1 | - | 1 - 2 | 71 | 101 | |
| DC422367.1 | - | 1 - 2 | 96 | 101 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000550.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK300427.1,NM_001136562.2 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000259318.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000259318.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000259318.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000259318.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000259318.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000259318.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000259318.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000259318.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000259318.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000259318.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000471798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000471798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000471798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000480388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000480388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000480388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000480388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000480388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000482335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000482335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000482335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000482335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000482335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000482335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000485762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000485762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000495980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000495980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000495980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | ENST00000495980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006135.hg.1 | ENST00000259318.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006135.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006135.hg.1 | TC09000550.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0900184402.hg.1 | ENST00000259318.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184402.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184402.hg.1 | ENST00000259318.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184402.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184402.hg.1 | TC0900012157.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000259318.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000259318.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000259318.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000259318.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000259318.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000259318.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000259318.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000259318.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000259318.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000259318.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000374525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000434623.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000471798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000471798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000471798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000480388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000480388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000480388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000480388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000480388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000482335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000482335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000482335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000482335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000482335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000482335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000485762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000485762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000495980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000495980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000495980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | ENST00000495980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001004065.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001136562.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001198656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012157.hg.1 | NM_001198656.1 | Refseq |

AKAP2
Genomic Map