Assay Details
| Gene Symbol: S100A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020672.2 | NP_065723.1 | 2 - 3 | 145 | 89 | |
| XM_005245362.1 | XP_005245419.1 | 2 - 3 | 141 | 89 | ||
| XM_017001875.1 | XP_016857364.1 | 3 - 4 | 233 | 89 | ||
| GenBank mRNA | AY007220.1 | - | 2 - 3 | 127 | 89 | |
| BC005019.1 | - | 1 - 2 | 109 | 3640899 | 89 | |
| BG677973.1 | - | 2 - 3 | 145 | 89 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003270.hg.1
Additional Information:
This assay is in the 5'UTR region of the AY007220.1,BC005019.1,NM_020672.2,XM_005245362.1,XM_017001875.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
response to lipopolysaccharide
toll-like receptor 4 signaling pathway
defense response to bacterium
calcium ion homeostasis
positive regulation of granulocyte chemotaxis
positive regulation of monocyte chemotaxis
calcium ion binding
protein binding
chemokine receptor binding
apoptotic process
response to lipopolysaccharide
toll-like receptor 4 signaling pathway
defense response to bacterium
calcium ion homeostasis
positive regulation of granulocyte chemotaxis
positive regulation of monocyte chemotaxis
calcium ion binding
protein binding
chemokine receptor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01050314.hg.1 | ENST00000344616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050314.hg.1 | ENST00000368700.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050314.hg.1 | ENST00000368701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050314.hg.1 | ENST00000368702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050314.hg.1 | ENST00000476873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050314.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050314.hg.1 | TC01003270.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050315.hg.1 | ENST00000344616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050315.hg.1 | ENST00000368701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050315.hg.1 | ENST00000368702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050315.hg.1 | ENST00000476873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050315.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050315.hg.1 | TC01003270.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050324.hg.1 | ENST00000368702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050324.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050324.hg.1 | TC01003270.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000469571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000469571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000469571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000469571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000469571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000476873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000476873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000476873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000476873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000476873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000476873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000476873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | ENST00000476873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015872.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050316.hg.1 | ENST00000344616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050316.hg.1 | ENST00000368700.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050316.hg.1 | ENST00000368701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050316.hg.1 | ENST00000368702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050316.hg.1 | ENST00000476873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050316.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050316.hg.1 | TC01003270.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100219246.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219246.hg.1 | ENST00000368700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219246.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219246.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219246.hg.1 | ENST00000469571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219246.hg.1 | ENST00000476873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219246.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219246.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219246.hg.1 | ENST00000368700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219246.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219246.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219246.hg.1 | ENST00000469571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219246.hg.1 | ENST00000476873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219246.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219246.hg.1 | TC0100015872.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100219253.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219253.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219253.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219253.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219253.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219253.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219253.hg.1 | TC0100015872.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100219247.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219247.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219247.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219247.hg.1 | ENST00000469571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219247.hg.1 | ENST00000476873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219247.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219247.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219247.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219247.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219247.hg.1 | ENST00000469571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219247.hg.1 | ENST00000476873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219247.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219247.hg.1 | TC0100015872.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100219249.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219249.hg.1 | ENST00000368700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219249.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219249.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219249.hg.1 | ENST00000469571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219249.hg.1 | ENST00000476873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219249.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219249.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219249.hg.1 | ENST00000368700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219249.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219249.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219249.hg.1 | ENST00000469571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219249.hg.1 | ENST00000476873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219249.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219249.hg.1 | TC0100015872.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000344616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000368702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000469571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000469571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000469571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000469571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000469571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000476873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000476873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000476873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000476873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000476873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000476873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000476873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | ENST00000476873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | NM_020672.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015872.hg.1 | NM_020672.2 | Refseq |

S100A14
Genomic Map