Assay Details
| Gene Symbol: ENOPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001292017.1 | NP_001278946.1 | 4 - 5 | 841 | 93 | |
| NM_021204.4 | NP_067027.1 | 4 - 5 | 870 | 93 | ||
| NR_120457.1 | 3 - 4 | 638 | 93 | |||
| GenBank mRNA | AF087881.1 | - | 4 - 5 | 740 | 93 | |
| AF113125.1 | - | 4 - 5 | 741 | 93 | ||
| AF177286.1 | - | 2 - 3 | 307 | 93 | ||
| AL518796.3 | - | 4 - 5 | 673 | 93 | ||
| BC001317.1 | - | 3 - 4 | 456 | 93 | ||
| BC065815.1 | - | 4 - 5 | 708 | 6526329 | 93 | |
| CR457141.1 | - | 4 - 5 | 520 | 93 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000454.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000505846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000505846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000505846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000505846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000505846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000505846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000505846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000505846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000509635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000509635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000509635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000509635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000509635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000509635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000509635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000509635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000509635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000509635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000610545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000610545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000610545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000610545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000610545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000610545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000610545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006505.hg.1 | ENST00000273920.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006505.hg.1 | ENST00000456931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006505.hg.1 | ENST00000509635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006505.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006505.hg.1 | TC04000454.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006506.hg.1 | ENST00000273920.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006506.hg.1 | ENST00000456931.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006506.hg.1 | ENST00000509635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006506.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006506.hg.1 | TC04000454.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000505846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000505846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000505846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000505846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000505846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000505846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000505846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000505846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000509635.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000509635.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000509635.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000509635.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000509635.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000509635.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000509635.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000509635.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000509635.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000509635.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000610545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000610545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000610545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000610545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000610545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000610545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000610545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007978.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400154900.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154900.hg.1 | ENST00000505846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154900.hg.1 | ENST00000509635.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154900.hg.1 | ENST00000610545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154900.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154900.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400154900.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400154900.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400154900.hg.1 | ENST00000273920.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154900.hg.1 | ENST00000505846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154900.hg.1 | ENST00000509635.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154900.hg.1 | ENST00000610545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154900.hg.1 | ENST00000618400.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154900.hg.1 | NM_001292017.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400154900.hg.1 | NM_021204.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400154900.hg.1 | NR_120457.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400154900.hg.1 | TC0400007978.hg.1 | Clariom D TC |

ENOPH1
Genomic Map