Assay Details
| Gene Symbol: GPR157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024980.4 | NP_079256.4 | 1 - 2 | 521 | 71 | |
| XM_005263496.4 | XP_005263553.1 | 1 - 2 | 470 | 71 | ||
| XM_005263497.4 | XP_005263554.1 | 1 - 2 | 470 | 71 | ||
| GenBank mRNA | AK292888.1 | - | 1 - 2 | 475 | 71 | |
| AK304154.1 | - | 1 - 2 | 475 | 71 | ||
| BC094733.1 | - | 3 - 4 | 368 | 71 | ||
More Information
Set Membership: |
|

GPR157
Genomic Map