| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03037584.hg.1 |
ENST00000347624.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03037584.hg.1 |
ENST00000428839.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03037584.hg.1 |
NM_001135057.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03037584.hg.1 |
NM_130830.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03037584.hg.1 |
TC03002131.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013598.hg.1 |
ENST00000347624.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013598.hg.1 |
ENST00000347624.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013598.hg.1 |
ENST00000347624.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013598.hg.1 |
ENST00000347624.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013598.hg.1 |
ENST00000428839.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013598.hg.1 |
ENST00000428839.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013598.hg.1 |
ENST00000428839.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013598.hg.1 |
ENST00000428839.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013598.hg.1 |
NM_001135057.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013598.hg.1 |
NM_001135057.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013598.hg.1 |
NM_001135057.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013598.hg.1 |
NM_001135057.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013598.hg.1 |
NM_001135057.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013598.hg.1 |
NM_130830.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013598.hg.1 |
NM_130830.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013598.hg.1 |
NM_130830.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013598.hg.1 |
NM_130830.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03037580.hg.1 |
ENST00000347624.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03037580.hg.1 |
ENST00000428839.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03037580.hg.1 |
ENST00000439944.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03037580.hg.1 |
NM_001135057.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03037580.hg.1 |
NM_130830.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03037580.hg.1 |
TC03002131.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300201439.hg.1 |
ENST00000347624.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300201439.hg.1 |
ENST00000428839.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300201439.hg.1 |
NM_001135057.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300201439.hg.1 |
NM_130830.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300201439.hg.1 |
ENST00000347624.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300201439.hg.1 |
ENST00000428839.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300201439.hg.1 |
NM_001135057.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300201439.hg.1 |
NM_130830.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300201439.hg.1 |
TC0300013598.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300201443.hg.1 |
ENST00000347624.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300201443.hg.1 |
ENST00000428839.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300201443.hg.1 |
NM_001135057.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300201443.hg.1 |
NM_130830.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300201443.hg.1 |
ENST00000347624.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300201443.hg.1 |
ENST00000428839.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300201443.hg.1 |
NM_001135057.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300201443.hg.1 |
NM_130830.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300201443.hg.1 |
TC0300013598.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300013598.hg.1 |
ENST00000347624.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013598.hg.1 |
ENST00000347624.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013598.hg.1 |
ENST00000347624.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013598.hg.1 |
ENST00000347624.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013598.hg.1 |
ENST00000428839.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013598.hg.1 |
ENST00000428839.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013598.hg.1 |
ENST00000428839.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013598.hg.1 |
ENST00000428839.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013598.hg.1 |
NM_001135057.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013598.hg.1 |
NM_001135057.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013598.hg.1 |
NM_001135057.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013598.hg.1 |
NM_001135057.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013598.hg.1 |
NM_001135057.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013598.hg.1 |
NM_130830.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013598.hg.1 |
NM_130830.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013598.hg.1 |
NM_130830.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013598.hg.1 |
NM_130830.4 |
Refseq |