| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002325.hg.1 |
ENST00000307471.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002325.hg.1 |
ENST00000409765.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002325.hg.1 |
ENST00000410069.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002325.hg.1 |
ENST00000486109.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002325.hg.1 |
NM_138771.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002325.hg.1 |
TC07000149.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002324.hg.1 |
ENST00000307471.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002324.hg.1 |
ENST00000409765.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002324.hg.1 |
ENST00000410069.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002324.hg.1 |
ENST00000486109.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002324.hg.1 |
NM_138771.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07002324.hg.1 |
TC07000149.hg.1 |
HTA TC |