Assay Details
| Gene Symbol: TMEM217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145316.3 | NP_660359.2 | 3 - 4 | 768 | 87 | |
| NR_104442.1 | 2 - 3 | 670 | 87 | |||
| XM_011514362.1 | XP_011512664.1 | 4 - 5 | 780 | 87 | ||
| XM_011514364.2 | XP_011512666.1 | 4 - 5 | 1564 | 87 | ||
| XM_011514365.2 | XP_011512667.1 | 4 - 5 | 1349 | 87 | ||
| GenBank mRNA | AK098666.1 | - | 3 - 4 | 727 | 87 | |
| DB458508.1 | - | 2 - 3 | 152 | 87 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001682.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06020063.hg.1 | ENST00000336655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020063.hg.1 | ENST00000356757.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020063.hg.1 | NM_001162900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020063.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020063.hg.1 | TC06001682.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020057.hg.1 | ENST00000336655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020057.hg.1 | ENST00000356757.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020057.hg.1 | ENST00000497775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020057.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020057.hg.1 | TC06001682.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020058.hg.1 | ENST00000336655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020058.hg.1 | ENST00000356757.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020058.hg.1 | ENST00000497775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020058.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020058.hg.1 | TC06001682.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000336655.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000336655.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000336655.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000336655.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000336655.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000336655.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000356757.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000356757.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000356757.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000356757.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000356757.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000356757.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000357219.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000357219.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000357219.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000357219.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000357219.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000478262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000478262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000478262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000478262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000482580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000482580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000497775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000497775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000497775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | ENST00000497775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NM_001162900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NM_001162900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NM_001162900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NM_001162900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NM_001162900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NM_001162900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NM_001162900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NM_001286401.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NM_001286401.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NM_001286401.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NM_001286401.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NM_001286401.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NM_001286401.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NM_001286401.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NR_104442.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NR_104442.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NR_104442.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NR_104442.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NR_104442.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NR_104442.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NR_104443.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NR_104443.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NR_104443.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NR_104443.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011690.hg.1 | NR_104443.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020059.hg.1 | ENST00000336655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020059.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020059.hg.1 | TC06001682.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000336655.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000336655.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000336655.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000336655.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000336655.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000336655.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000356757.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000356757.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000356757.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000356757.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000356757.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000356757.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000357219.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000357219.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000357219.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000357219.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000357219.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000478262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000478262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000478262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000478262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000482580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000482580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000497775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000497775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000497775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | ENST00000497775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NM_001162900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NM_001162900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NM_001162900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NM_001162900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NM_001162900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NM_001162900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NM_001162900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NM_001286401.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NM_001286401.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NM_001286401.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NM_001286401.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NM_001286401.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NM_001286401.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NM_001286401.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NR_104442.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NR_104442.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NR_104442.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NR_104442.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NR_104442.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NR_104442.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NR_104443.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NR_104443.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NR_104443.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NR_104443.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011690.hg.1 | NR_104443.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600180264.hg.1 | ENST00000336655.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600180264.hg.1 | ENST00000356757.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600180264.hg.1 | ENST00000357219.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600180264.hg.1 | ENST00000482580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600180264.hg.1 | NM_001162900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600180264.hg.1 | NM_001286401.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600180264.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600180264.hg.1 | ENST00000336655.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600180264.hg.1 | ENST00000356757.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600180264.hg.1 | ENST00000357219.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600180264.hg.1 | ENST00000482580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600180264.hg.1 | NM_001162900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600180264.hg.1 | NM_001286401.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600180264.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600180264.hg.1 | TC0600011690.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600180259.hg.1 | ENST00000336655.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600180259.hg.1 | ENST00000356757.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600180259.hg.1 | ENST00000478262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600180259.hg.1 | ENST00000497775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600180259.hg.1 | NM_001286401.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600180259.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600180259.hg.1 | NR_104442.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600180259.hg.1 | NR_104443.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600180259.hg.1 | ENST00000336655.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600180259.hg.1 | ENST00000356757.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600180259.hg.1 | ENST00000478262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600180259.hg.1 | ENST00000497775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600180259.hg.1 | NM_001286401.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600180259.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600180259.hg.1 | NR_104442.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600180259.hg.1 | NR_104443.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600180259.hg.1 | TC0600011690.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600180260.hg.1 | ENST00000336655.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600180260.hg.1 | ENST00000478262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600180260.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600180260.hg.1 | NR_104442.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600180260.hg.1 | ENST00000336655.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600180260.hg.1 | ENST00000478262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600180260.hg.1 | NM_145316.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600180260.hg.1 | NR_104442.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600180260.hg.1 | TC0600011690.hg.1 | Clariom D TC |

TMEM217
Genomic Map