Assay Details
| Gene Symbol: TBC1D3F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK292344.1 | - | 9 - 10 | 825 | 107 | |
| BC075809.1 | - | 9 - 10 | 814 | 5170064 | 107 | |
| Gene Symbol: TBC1D3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001417.6 | NP_001001417.6 | 9 - 10 | 814 | 107 | |
| XM_005257980.4 | XP_005258037.2 | 9 - 10 | 944 | 107 | ||
| GenBank mRNA | AK292257.1 | - | 9 - 10 | 785 | 107 | |
| Gene Symbol: TBC1D3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK292308.1 | - | 10 - 11 | 818 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

TBC1D3F
Genomic Map