Assay Details
| Gene Symbol: PRMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001207042.2 | NP_001193971.1 | 1 - 2 | 169 | 95 | |
| NM_198318.4 | NP_938074.2 | 1 - 2 | 169 | 95 | ||
| GenBank mRNA | AK304660.1 | - | 1 - 2 | 169 | 95 | |
| DC421815.1 | - | 1 - 2 | 169 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK304660.1,NM_001207042.2,NM_198318.4 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

PRMT1
Genomic Map