| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011687.hg.1 |
ENST00000490933.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01026068.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01026068.hg.1 |
ENST00000468449.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01026068.hg.1 |
NM_001079809.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01026068.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01026068.hg.1 |
TC01001681.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007469.hg.1 |
ENST00000432739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000463015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000463015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000463015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000468449.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000468449.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000468449.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000468449.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000468449.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000468449.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000468449.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000488097.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000488097.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000488097.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000488097.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000488097.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000543891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000543891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000543891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000543891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000543891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000543891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000543891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
ENST00000543891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01026066.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01026066.hg.1 |
NM_001079809.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01026066.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01026066.hg.1 |
TC01001681.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008890.hg.1 |
ENST00000561684.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007074.hg.1 |
ENST00000503901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012790.hg.1 |
ENST00000463122.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012790.hg.1 |
NM_001079809.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012790.hg.1 |
NM_001079809.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012790.hg.1 |
NM_001079809.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100182490.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100182490.hg.1 |
ENST00000543891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100182490.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100182490.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100182490.hg.1 |
ENST00000543891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100182490.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100182490.hg.1 |
TC0100011242.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000463015.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000463015.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000463015.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000468449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000468449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000468449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000468449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000468449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000468449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000468449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000488097.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000488097.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000488097.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000488097.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000488097.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000543891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000543891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000543891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000543891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000543891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000543891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000543891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
ENST00000543891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011242.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011687.hg.1 |
ENST00000490933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008890.hg.1 |
ENST00000561684.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100182493.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100182493.hg.1 |
ENST00000468449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100182493.hg.1 |
ENST00000543891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100182493.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100182493.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100182493.hg.1 |
ENST00000468449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100182493.hg.1 |
ENST00000543891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100182493.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100182493.hg.1 |
TC0100011242.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100182491.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100182491.hg.1 |
ENST00000543891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100182491.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100182491.hg.1 |
ENST00000367242.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100182491.hg.1 |
ENST00000543891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100182491.hg.1 |
NM_138391.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100182491.hg.1 |
TC0100011242.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300012790.hg.1 |
ENST00000463122.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012790.hg.1 |
NM_001079809.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012790.hg.1 |
NM_001079809.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012790.hg.1 |
NM_001079809.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300195697.hg.1 |
ENST00000463122.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300195697.hg.1 |
NM_001079809.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300195697.hg.1 |
ENST00000463122.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300195697.hg.1 |
NM_001079809.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300195697.hg.1 |
TC0300012790.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600007469.hg.1 |
ENST00000432739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300195698.hg.1 |
NM_001079809.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300195698.hg.1 |
NM_001079809.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300195698.hg.1 |
TC0300012790.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500007074.hg.1 |
ENST00000503901.1 |
Ensembl |