Assay Details
| Gene Symbol: HS3ST3B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006041.2 | NP_006032.1 | 1 | 789 | 110 | |
| NR_130138.1 | 1 | 789 | 110 | |||
| GenBank mRNA | AF105377.1 | - | 1 | 650 | 110 | |
| AK023723.1 | - | 1 | 756 | 110 | ||
| BC063301.1 | - | 1 | 706 | 30343352 | 110 | |
| BC069664.1 | - | 1 | 590 | 7262483 | 110 | |
| BC069725.1 | - | 1 | 589 | 7262495 | 110 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000165.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
glycosaminoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process, enzymatic modification
[heparan sulfate]-glucosamine 3-sulfotransferase 1 activity
[heparan sulfate]-glucosamine 3-sulfotransferase 3 activity
glycosaminoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process, enzymatic modification
[heparan sulfate]-glucosamine 3-sulfotransferase 1 activity
[heparan sulfate]-glucosamine 3-sulfotransferase 3 activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17002300.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002300.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002300.hg.1 | TC17000165.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002303.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002303.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002303.hg.1 | TC17000165.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002302.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002302.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002302.hg.1 | TC17000165.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002299.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002299.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002299.hg.1 | TC17000165.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700148935.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148935.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148935.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148935.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148935.hg.1 | TC1700006946.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700148935.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148935.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148935.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148935.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148936.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148936.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148936.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148936.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148936.hg.1 | TC1700006946.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700148936.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148936.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148936.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148936.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148937.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148937.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148937.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148937.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148937.hg.1 | TC1700006946.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700148937.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148937.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148937.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148937.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148939.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148939.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148939.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148939.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148939.hg.1 | TC1700006946.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700148939.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148939.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148939.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148939.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148933.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148933.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148933.hg.1 | TC1700006946.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700148933.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148933.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148938.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148938.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148938.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148938.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148938.hg.1 | TC1700006946.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700148938.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148938.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148938.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148938.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148934.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148934.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148934.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148934.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148934.hg.1 | TC1700006946.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700148934.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148934.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148934.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148934.hg.1 | NR_130138.1 | Refseq |

HS3ST3B1
Genomic Map