| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013006.hg.1 |
ENST00000361987.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013006.hg.1 |
ENST00000361987.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013006.hg.1 |
ENST00000361987.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013006.hg.1 |
ENST00000361987.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013006.hg.1 |
ENST00000361987.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013006.hg.1 |
ENST00000361987.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013006.hg.1 |
NM_000614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013006.hg.1 |
NM_000614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013006.hg.1 |
NM_000614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013006.hg.1 |
NM_000614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013006.hg.1 |
NM_000614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013006.hg.1 |
NM_000614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11005806.hg.1 |
ENST00000361987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11005806.hg.1 |
NM_000614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11005806.hg.1 |
TC11000477.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1100200545.hg.1 |
ENST00000361987.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100200545.hg.1 |
NM_000614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100200545.hg.1 |
ENST00000361987.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100200545.hg.1 |
NM_000614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100200545.hg.1 |
TC1100013006.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100200547.hg.1 |
ENST00000361987.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100200547.hg.1 |
NM_000614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100200547.hg.1 |
ENST00000361987.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100200547.hg.1 |
NM_000614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100200547.hg.1 |
TC1100013006.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100013006.hg.1 |
ENST00000361987.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013006.hg.1 |
ENST00000361987.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013006.hg.1 |
ENST00000361987.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013006.hg.1 |
ENST00000361987.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013006.hg.1 |
ENST00000361987.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013006.hg.1 |
ENST00000361987.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013006.hg.1 |
NM_000614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013006.hg.1 |
NM_000614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013006.hg.1 |
NM_000614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013006.hg.1 |
NM_000614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013006.hg.1 |
NM_000614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013006.hg.1 |
NM_000614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100200546.hg.1 |
ENST00000361987.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100200546.hg.1 |
NM_000614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100200546.hg.1 |
ENST00000361987.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100200546.hg.1 |
NM_000614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100200546.hg.1 |
TC1100013006.hg.1 |
Clariom D TC |