Assay Details
| Gene Symbol: NUDT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301022.1 | NP_001287951.1 | 5 | 4091 | 103 | |
| NM_001301023.1 | NP_001287952.1 | 5 | 4088 | 103 | ||
| NM_001301024.1 | NP_001287953.1 | 5 | 3991 | 103 | ||
| NM_019094.5 | NP_061967.3 | 5 | 4346 | 103 | ||
| NM_199040.3 | NP_950241.1 | 5 | 4349 | 103 | ||
| GenBank mRNA | AF191651.1 | - | 5 | 3934 | 103 | |
| AF191652.2 | - | 5 | 3935 | 103 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NUDT4
Genomic Map