Assay Details
| Gene Symbol: MIRLET7A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029477.1 | 1 | 36 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
BH3-like motif containing, cell death inducer |
BLID | BRCC2 | -30.3kb | Sense | Chr. 11 - 122115354 - 122116215 on NCBI Build GRCh38 |
| Downstream | microRNA 100 | MIR100 |
MIRN100, miR-100 |
5.6kb | Sense | Chr. 11 - 122152229 - 122152308 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1100012588.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11002384.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08081247 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508774 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508810 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508811 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508775 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08081263 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508804 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508785 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08081234 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08081287 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508795 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508772 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508815 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508791 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508813 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508788 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508807 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508809 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08081253 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08081241 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08081240 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08081260 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508781 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508799 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508817 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08081286 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508806 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08081243 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508800 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508811 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08081260 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508807 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508813 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508772 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08081243 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508800 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508785 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508795 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508799 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508817 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508781 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08081263 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508804 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08081286 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508775 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508774 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508788 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508809 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08081241 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508810 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08081287 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508815 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08081253 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508806 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08081234 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08081240 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28508791 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08081247 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.1 ST Array | 7952313 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR11028686.hg.1 | ENST00000362105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11028686.hg.1 | NR_029477.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11028686.hg.1 | TC11002384.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012588.hg.1 | ENST00000362105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012588.hg.1 | NR_029477.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012588.hg.1 | ENST00000362105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012588.hg.1 | NR_029477.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197057.hg.1 | ENST00000362105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197057.hg.1 | NR_029477.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197057.hg.1 | ENST00000362105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197057.hg.1 | NR_029477.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197057.hg.1 | TC1100012588.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3310131 | NR_029477.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3310131 | NR_029477.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3310131 | NR_029477.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3310131 | NR_029477.1 | human_transcriptome |

MIRLET7A2
Genomic Map