Assay Details
| Gene Symbol: MIR24-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029497.1 | 1 | 31 | 61 | ||
| GenBank mRNA | AF480559.1 | - | 1 | 38 | 61 | |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1900011896.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19001224.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Chr array_18 19 21 | A_17_P17111366 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17111369 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10890753 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10890749 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10890754 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10890750 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31506867 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17111370 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10890752 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31506862 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10890751 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10890755 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31506865 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31506866 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31506868 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10890752 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10890753 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10890751 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31506865 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10890755 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31506866 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17111370 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31506868 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31506862 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10890751 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10890753 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17111369 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31506867 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10890749 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31506865 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17111366 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17111370 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10890750 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10890752 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10890755 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31506868 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10890754 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31506866 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10890751 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10890752 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31506868 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17111370 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10890755 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31506866 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10890753 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31506865 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8034695 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8034694 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011896.hg.1 | ENST00000386972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011896.hg.1 | ENST00000587762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011896.hg.1 | NR_029497.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011896.hg.1 | NR_036515.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017209.hg.1 | ENST00000386972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017209.hg.1 | NR_029497.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017209.hg.1 | TC19001224.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900011896.hg.1 | ENST00000386972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011896.hg.1 | ENST00000587762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011896.hg.1 | NR_029497.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011896.hg.1 | NR_036515.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900204928.hg.1 | ENST00000386972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900204928.hg.1 | NR_029497.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900204928.hg.1 | TC1900011896.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900204928.hg.1 | ENST00000386972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900204928.hg.1 | NR_029497.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3310573 | NR_029497.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3310573 | AF480559.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3310573 | NR_029497.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3310573 | AF480559.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3310573 | NR_029497.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3310573 | AF480559.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3310573 | NR_029497.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3310573 | AF480559.1 | human_transcriptome |

MIR24-2
Genomic Map