Assay Details
| Gene Symbol: MIR25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029498.1 | 1 | 41 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | COP9 signalosome subunit 6 | COPS6 |
CSN6, MOV34-34KD |
-1.4kb | Anti-sense | Chr. 7 - 100088960 - 100092200 on NCBI Build GRCh38 |
| Downstream | microRNA 93 | MIR93 |
MIRN9, MIRN93, hsa-mir-93, miR-93 |
125.0 | Sense | Chr. 7 - 100093768 - 100093847 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0700011970.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07001655.hg.1
Set Membership: |
|

MIR25
Genomic Map