Assay Details
| Gene Symbol: MIR26A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029499.1 | 1 | 33 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ITGA9 antisense RNA 1 | ITGA9-AS1 | -107.6kb | Anti-sense | Chr. 3 - 37753689 - 37861780 on NCBI Build GRCh38 | |
| Downstream | villin like | VILL | 18.5kb | Sense | Chr. 3 - 37987974 - 38007188 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0300007062.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000183.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178070 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178051 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178069 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311725 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178118 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178089 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178079 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311747 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311737 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311690 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178059 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311698 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311726 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178066 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178074 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311679 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178053 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311728 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15428267 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311710 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178091 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311682 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311677 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178060 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311722 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311705 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311681 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178113 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311739 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311738 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178101 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311724 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178086 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311700 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311707 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311686 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178115 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311714 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311697 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178117 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178105 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178081 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178056 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311693 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178103 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178040 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311690 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178086 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178113 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178051 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311728 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15428267 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311739 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311693 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178105 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178103 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178079 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311698 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178091 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311738 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311705 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178089 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311700 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178056 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178040 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178117 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311725 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311679 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178066 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178118 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311707 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311677 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311686 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178053 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178101 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311714 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311747 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178069 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311682 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311710 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311737 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178081 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178115 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311722 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178059 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311726 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311724 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178070 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311681 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22311697 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178074 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02178060 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.1 ST Array | 8078663 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007062.hg.1 | ENST00000362205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007062.hg.1 | NR_029499.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03003483.hg.1 | ENST00000362205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03003483.hg.1 | NR_029499.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03003483.hg.1 | TC03000183.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0300007062.hg.1 | ENST00000362205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007062.hg.1 | NR_029499.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300149278.hg.1 | ENST00000362205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300149278.hg.1 | NR_029499.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300149278.hg.1 | ENST00000362205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300149278.hg.1 | NR_029499.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300149278.hg.1 | TC0300007062.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3310985 | NR_029499.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3310985 | NR_029499.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3310985 | NR_029499.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3310985 | NR_029499.1 | human_transcriptome |

MIR26A1
Genomic Map