Assay Details
| Gene Symbol: MIR33A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029507.1 | 1 | 26 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105373045 | LOC105373045 | -8.5kb | Sense | Chr. 22 - 41885186 - 41892439 on NCBI Build GRCh38 | |
| Downstream | shisa family member 8 | SHISA8 |
C22orf17, Orf26 |
8.5kb | Anti-sense | Chr. 22 - 41907180 - 41915093 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC2200007499.hg.1
This assay targets the same location as HTA transcript cluster(s) TC22000348.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513649 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513656 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513644 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200048 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513630 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200068 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513639 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513657 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513654 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286632 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513623 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200063 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513647 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200046 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286637 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286646 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200080 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200066 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513653 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286634 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513627 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286640 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200056 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286639 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513611 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286635 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200061 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513631 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286638 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513614 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513624 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513621 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200099 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513646 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200078 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513618 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513652 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513659 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513635 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286631 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200053 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286641 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200065 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286643 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286636 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513655 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513612 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286641 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513627 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286643 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200053 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200078 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513631 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513646 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200065 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513611 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513655 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286632 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513656 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513652 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513649 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200063 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513612 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200061 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513647 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200099 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286635 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286631 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513653 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513635 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200048 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513621 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286639 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200046 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513657 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200066 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200080 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513644 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286636 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286646 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513659 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513639 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513623 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286640 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200068 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32200056 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286634 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286637 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513624 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513614 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513630 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513654 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11513618 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17286638 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.1 ST Array | 8073544 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006746.hg.1 | ENST00000385197.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006746.hg.1 | NR_029507.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006746.hg.1 | TC22000348.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007499.hg.1 | ENST00000385197.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007499.hg.1 | NR_029507.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007499.hg.1 | ENST00000385197.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007499.hg.1 | NR_029507.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200153095.hg.1 | ENST00000385197.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200153095.hg.1 | NR_029507.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200153095.hg.1 | TC2200007499.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200153095.hg.1 | ENST00000385197.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200153095.hg.1 | NR_029507.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3308255 | NR_029507.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3308255 | NR_029507.1 | human_transcriptome |

MIR33A
Genomic Map