Assay Details
| Gene Symbol: MIR30D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029599.1 | 1 | 32 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102723694 | LOC102723694 | Sense | Chr. 8 - 134789112 - 134832091 on NCBI Build GRCh38 | ||
| Downstream | non-protein coding RNA 250 | NCRNA00250 | C8orf24 | 33.1kb | Anti-sense | Chr. 8 - 134838069 - 134842637 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0800011911.hg.1
This assay targets the same location as HTA transcript cluster(s) TC08001667.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775664 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775648 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775639 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775676 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775680 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775629 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775671 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775673 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775655 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775627 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775670 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775662 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493217 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775633 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493221 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775626 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493211 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775644 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493239 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775637 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775674 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493237 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493212 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493258 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775652 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775663 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493250 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775667 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775679 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493235 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775635 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775666 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775660 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493245 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775647 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775650 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775646 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775624 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493226 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493261 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775681 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775669 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775641 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775639 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493217 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775662 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493258 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775637 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775664 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775660 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493245 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775627 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775650 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775626 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493261 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493226 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775646 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493211 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775641 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775666 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775670 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775673 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775629 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775680 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775624 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493237 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775652 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775663 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493221 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493235 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775633 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775647 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775669 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775635 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775644 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775648 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775674 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775655 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493239 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775667 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775681 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775671 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493250 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775679 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26775676 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06493212 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.1 ST Array | 8153067 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011911.hg.1 | ENST00000362283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011911.hg.1 | NR_029599.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08021794.hg.1 | ENST00000362283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08021794.hg.1 | NR_029599.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08021794.hg.1 | TC08001667.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0800181030.hg.1 | ENST00000362283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800181030.hg.1 | NR_029599.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800181030.hg.1 | ENST00000362283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800181030.hg.1 | NR_029599.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800181030.hg.1 | TC0800011911.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800011911.hg.1 | ENST00000362283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011911.hg.1 | NR_029599.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3308996 | NR_029599.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3308996 | NR_029599.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3308996 | NR_029599.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3308996 | NR_029599.1 | human_transcriptome |

MIR30D
Genomic Map