Assay Details
| Gene Symbol: MIR10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029609.1 | 1 | 53 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | HOXD cluster antisense RNA 2 | HOXD-AS2 | -13.2kb | Anti-sense | Chr. 2 - 176134841 - 176137098 on NCBI Build GRCh38 | |
| Downstream | HOXD antisense growth-associated long non-coding RNA | HAGLR |
HOXD-AS1, Mdgt |
22.8kb | Anti-sense | Chr. 2 - 176173189 - 176188958 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0200010051.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001054.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695514 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819725 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695481 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819672 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695513 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695486 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695505 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695524 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695502 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819728 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695509 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695471 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819664 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695454 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819679 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819698 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695504 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695457 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695467 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695496 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819694 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695526 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819696 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819716 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819682 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695464 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819681 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695525 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819726 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819659 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819690 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695520 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695484 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695511 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695523 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695510 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819667 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695516 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695521 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695460 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695522 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695456 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819676 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819686 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695493 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819692 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695506 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695507 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819700 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695526 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695486 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695484 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695454 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819676 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819659 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695460 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695514 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695502 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819725 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695471 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819692 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819700 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819667 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695521 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695506 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695464 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695456 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695523 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695496 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695493 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695457 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819716 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695525 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819728 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695504 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819686 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695520 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695467 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695516 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819664 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695513 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695510 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695507 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819726 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819679 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819694 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695509 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695505 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819690 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695481 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819698 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695511 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819681 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695524 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819696 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01695522 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819682 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21819672 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.1 ST Array | 8046553 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010051.hg.1 | ENST00000385011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010051.hg.1 | NR_029609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02015681.hg.1 | ENST00000385011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02015681.hg.1 | ENST00000549582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02015681.hg.1 | NR_029609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02015681.hg.1 | TC02001054.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200170538.hg.1 | ENST00000385011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200170538.hg.1 | NR_029609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200170538.hg.1 | ENST00000385011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200170538.hg.1 | NR_029609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200170538.hg.1 | TC0200010051.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200010051.hg.1 | ENST00000385011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010051.hg.1 | NR_029609.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A016634 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A016634 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3309484 | NR_029609.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3309484 | NR_029609.1 | human_transcriptome |

MIR10B
Genomic Map