Assay Details
| Gene Symbol: MIR218-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029632.1 | 1 | 55 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927969 | LOC101927969 | -47.3kb | Anti-sense | Chr. 5 - 168706927 - 168720884 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC728095 | LOC728095 | 245.0kb | Anti-sense | Chr. 5 - 169013227 - 169037998 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0500012762.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05002044.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451814 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630855 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451843 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451804 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630816 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451866 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451839 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451862 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451803 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451811 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451819 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630843 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451837 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451832 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451800 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451807 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451809 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451835 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451868 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451847 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451799 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451808 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451824 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15761313 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451829 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451870 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451817 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630810 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630834 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630837 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451863 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630823 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451864 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630836 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451841 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630832 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630821 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451849 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451805 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451851 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451867 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451854 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630841 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451807 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451851 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630821 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451817 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451799 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451837 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451870 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451803 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451808 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451841 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451832 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451862 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451809 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451843 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451829 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451864 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630834 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451814 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630843 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451866 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451805 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630816 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451804 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630823 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451863 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451819 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451839 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451800 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451847 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630836 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630837 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630832 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630841 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630810 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451849 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451867 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24630855 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451811 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451824 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451868 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451835 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04451854 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15761313 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.1 ST Array | 8115730 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012762.hg.1 | ENST00000385006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012762.hg.1 | NR_029632.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05028197.hg.1 | ENST00000385006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05028197.hg.1 | NR_029632.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05028197.hg.1 | TC05002044.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0500188916.hg.1 | ENST00000385006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500188916.hg.1 | NR_029632.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500188916.hg.1 | ENST00000385006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500188916.hg.1 | NR_029632.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500188916.hg.1 | TC0500012762.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500012762.hg.1 | ENST00000385006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012762.hg.1 | NR_029632.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3310376 | NR_029632.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3310376 | NR_029632.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3310376 | NR_029632.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3310376 | NR_029632.1 | human_transcriptome |

MIR218-2
Genomic Map