Assay Details
| Gene Symbol: MIR153-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029689.1 | 1 | 47 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100506557 | LOC100506557 | -47.0kb | Anti-sense | Chr. 7 - 157525740 - 157527319 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC105375614 | LOC105375614 | 39.6kb | Anti-sense | Chr. 7 - 157614023 - 157618771 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0700013255.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07002078.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167491 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898181 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167494 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898187 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167512 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898215 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898182 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167489 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167485 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898212 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898169 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898183 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898210 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167521 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167520 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167504 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16047119 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898213 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898193 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898197 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167517 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167522 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167483 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167509 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167530 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167526 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167492 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167528 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898186 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16047115 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898198 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167495 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898194 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16047136 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167529 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16047122 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167508 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167488 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167517 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167495 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167520 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167509 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898194 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167485 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898197 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898182 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898186 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167483 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16047115 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167526 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16047119 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167504 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167512 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167521 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898183 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167528 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167530 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16047136 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898212 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167488 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898187 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898215 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167489 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898193 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898213 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167492 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898169 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16047122 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167529 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898198 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167508 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167491 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167494 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26167522 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898210 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05898181 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.1 ST Array | 8144149 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR07031742.hg.1 | ENST00000385225.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07031742.hg.1 | NR_029689.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07031742.hg.1 | TC07002078.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013255.hg.1 | ENST00000385225.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013255.hg.1 | NR_029689.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013255.hg.1 | ENST00000385225.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013255.hg.1 | NR_029689.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700195514.hg.1 | ENST00000385225.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700195514.hg.1 | NR_029689.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700195514.hg.1 | ENST00000385225.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700195514.hg.1 | NR_029689.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700195514.hg.1 | TC0700013255.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3309384 | NR_029689.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3309384 | NR_029689.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309384 | NR_029689.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309384 | NR_029689.1 | human_transcriptome |

MIR153-2
Genomic Map