Assay Details
| Gene Symbol: MIR26A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029847.1 | 1 | 44 | 82 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | advillin | AVIL |
ADVIL, DOC6, p92 |
-4.8kb | Sense | Chr. 12 - 57797372 - 57818704 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105369783 | LOC105369783 | 1.7kb | Anti-sense | Chr. 12 - 57836625 - 57843761 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1200010985.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12001638.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379395 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379414 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814840 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379397 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814829 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16563651 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814831 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379379 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379385 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379388 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379393 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379410 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814833 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379402 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379382 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814825 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379375 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814871 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814878 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379364 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379384 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379399 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379377 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379362 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814873 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814862 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379369 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379365 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379416 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379407 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379409 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379412 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379386 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814846 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379408 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814849 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814863 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379380 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379361 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379388 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379393 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379414 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814829 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814871 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379395 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379384 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379365 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379399 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379407 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814833 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814825 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379362 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379409 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379408 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379377 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379369 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379385 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379412 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379380 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379416 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379382 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814878 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379379 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379364 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814840 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379375 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814846 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814862 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379410 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814863 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814831 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379386 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379402 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16563651 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379397 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814873 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28814849 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08379361 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.1 ST Array | 7964592 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR12022184.hg.1 | ENST00000385054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12022184.hg.1 | NR_029847.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12022184.hg.1 | TC12001638.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010985.hg.1 | ENST00000385054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010985.hg.1 | NR_029847.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200185646.hg.1 | ENST00000385054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200185646.hg.1 | NR_029847.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200185646.hg.1 | ENST00000385054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200185646.hg.1 | NR_029847.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200185646.hg.1 | TC1200010985.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200010985.hg.1 | ENST00000385054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010985.hg.1 | NR_029847.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3308981 | NR_029847.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3308981 | NR_029847.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3308981 | NR_029847.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3308981 | NR_029847.1 | human_transcriptome |

MIR26A2
Genomic Map