Assay Details
| Gene Symbol: MIR135B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029893.1 | 1 | 49 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | bladder cancer associated transcript 1 (non-protein coding) | BLACAT1 |
LINC00912, linc-UBC1, onco-lncRNA-30 |
Sense | Chr. 1 - 205434886 - 205456086 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC284577 | LOC284577 | 7.5kb | Anti-sense | Chr. 1 - 205456784 - 205469106 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0100017068.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01003748.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20808877 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747395 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747370 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747407 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20808903 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15166258 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15166238 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20808886 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747385 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20808853 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747405 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747353 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20808862 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747397 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747356 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747386 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747380 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747341 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747412 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747402 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747401 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747379 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747359 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20808872 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15166241 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747398 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747414 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747371 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747376 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747346 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747350 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747368 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20808893 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747340 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747416 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747392 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747404 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747365 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747394 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747388 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747344 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747338 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747383 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20808838 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747367 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747406 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747391 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747378 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747361 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20808890 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15166253 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747373 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20808829 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15166266 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15166261 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747342 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20808824 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747362 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747419 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15166267 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747392 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747342 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747412 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747373 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20808903 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747379 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747341 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747344 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20808862 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20808877 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747359 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747404 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747365 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747388 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20808829 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20808853 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747368 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747395 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747383 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20808890 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747419 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747361 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20808893 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747385 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15166241 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747346 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747394 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747405 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747406 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747401 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747391 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747370 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747386 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747416 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15166261 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747338 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747353 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15166253 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747362 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20808872 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747397 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747356 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747371 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20808838 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747414 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747367 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747340 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15166266 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747402 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15166238 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20808886 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747380 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747376 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747398 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15166258 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747350 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747378 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20808824 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00747407 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15166267 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01057492.hg.1 | ENST00000362189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057492.hg.1 | NR_029893.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057492.hg.1 | TC01003748.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017068.hg.1 | ENST00000362189.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017068.hg.1 | NR_029893.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229321.hg.1 | ENST00000362189.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229321.hg.1 | NR_029893.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229321.hg.1 | ENST00000362189.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229321.hg.1 | NR_029893.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229321.hg.1 | TC0100017068.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100017068.hg.1 | ENST00000362189.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017068.hg.1 | NR_029893.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3308976 | NR_029893.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3308976 | NR_029893.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3308976 | NR_029893.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3308976 | NR_029893.1 | human_transcriptome |

MIR135B
Genomic Map