Assay Details
| Gene Symbol: MIR422A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029944.1 | 1 | 41 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | HECT and RLD domain containing E3 ubiquitin protein ligase family member 1 | HERC1 |
p532, p619 |
-37.0kb | Sense | Chr. 15 - 63608618 - 63833964 on NCBI Build GRCh38 |
| Downstream | death associated protein kinase 2 | DAPK2 |
DRP-1, DRP1 |
36.0kb | Sense | Chr. 15 - 63907036 - 64046470 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1500009724.hg.1
Set Membership: |
|

MIR422A
Genomic Map