Assay Details
| Gene Symbol: MIR202 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030170.1 | 1 | 56 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MIR202 host gene | MIR202HG | Sense | Chr. 10 - 133246479 - 133247885 on NCBI Build GRCh38 | ||
| Downstream | ADAM metallopeptidase domain 8 | ADAM8 |
CD156, CD156a, MS2 |
14.8kb | Sense | Chr. 10 - 133262416 - 133276903 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1000012387.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001782.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592631 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592617 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592633 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592655 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592650 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592616 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592634 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16403428 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592629 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592619 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592624 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27984945 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592648 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592623 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592652 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592625 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592653 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592646 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592658 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592641 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592657 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592618 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592639 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592647 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16403432 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592642 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16403425 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592654 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592630 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27984940 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27984997 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592635 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592644 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592638 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27984958 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592632 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592637 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592626 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27984946 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592651 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27984974 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592619 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592652 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592647 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592634 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592629 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592623 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592639 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592650 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592633 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592624 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592638 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592644 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592616 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592654 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592630 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592641 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592657 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592626 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592637 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592642 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16403428 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592651 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592635 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27984945 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27984940 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27984958 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592625 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16403432 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592646 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16403425 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592648 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27984997 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27984946 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592631 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592655 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592617 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592653 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592658 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592632 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592618 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27984974 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.1 ST Array | 7937148 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023499.hg.1 | ENST00000300167.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023499.hg.1 | ENST00000362219.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023499.hg.1 | NR_030170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023499.hg.1 | TC10001782.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012387.hg.1 | ENST00000300167.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012387.hg.1 | ENST00000300167.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012387.hg.1 | ENST00000300167.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012387.hg.1 | ENST00000300167.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012387.hg.1 | ENST00000362219.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012387.hg.1 | ENST00000362219.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012387.hg.1 | ENST00000553459.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012387.hg.1 | ENST00000553459.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012387.hg.1 | ENST00000553459.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012387.hg.1 | NR_030170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012387.hg.1 | NR_030170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012387.hg.1 | NR_108078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012387.hg.1 | NR_108078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012387.hg.1 | NR_108078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012387.hg.1 | NR_108078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012387.hg.1 | NR_108078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012387.hg.1 | NR_108079.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012387.hg.1 | NR_108079.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012387.hg.1 | NR_108079.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023498.hg.1 | ENST00000362219.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023498.hg.1 | NR_030170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023498.hg.1 | TC10001782.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000012387.hg.1 | ENST00000300167.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012387.hg.1 | ENST00000300167.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012387.hg.1 | ENST00000300167.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012387.hg.1 | ENST00000300167.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012387.hg.1 | ENST00000362219.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012387.hg.1 | ENST00000362219.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012387.hg.1 | ENST00000553459.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012387.hg.1 | ENST00000553459.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012387.hg.1 | ENST00000553459.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012387.hg.1 | NR_030170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012387.hg.1 | NR_030170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012387.hg.1 | NR_108078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012387.hg.1 | NR_108078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012387.hg.1 | NR_108078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012387.hg.1 | NR_108078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012387.hg.1 | NR_108078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012387.hg.1 | NR_108079.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012387.hg.1 | NR_108079.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012387.hg.1 | NR_108079.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184487.hg.1 | ENST00000362219.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184487.hg.1 | NR_030170.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184487.hg.1 | ENST00000362219.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184487.hg.1 | NR_030170.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184487.hg.1 | TC1000012387.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3310920 | NR_030170.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3310920 | NR_030170.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3310920 | NR_030170.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3310920 | NR_030170.1 | human_transcriptome |

MIR202
Genomic Map