Assay Details
| Gene Symbol: MIR193B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030177.1 | 1 | 46 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MKL1/myocardin like 2 | MKL2 |
MRTF-B, MRTFB, NPD001 |
-37.2kb | Sense | Chr. 16 - 14071076 - 14266779 on NCBI Build GRCh38 |
| Downstream | microRNA 365a | MIR365A |
MIR365-1, MIRN365-1, hsa-mir-365a, mir-365a |
5.2kb | Sense | Chr. 16 - 14309285 - 14309371 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1600006986.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000174.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939196 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451627 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939204 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939185 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939202 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939198 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451623 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939194 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451628 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939171 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451651 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939170 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939184 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939174 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451642 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939182 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939190 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451631 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853239 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939176 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939186 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853240 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939179 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451639 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939192 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451632 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853237 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451625 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451624 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451622 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939169 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853242 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451657 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451624 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451651 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939169 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939198 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939192 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939196 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939186 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939194 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853240 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939174 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451632 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939171 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853242 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451627 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451639 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451628 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939184 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939204 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939176 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939185 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451622 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451625 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939170 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451642 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853239 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853237 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451623 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451631 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939179 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939190 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939182 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451657 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939202 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600006986.hg.1 | ENST00000384907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006986.hg.1 | NR_030177.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002632.hg.1 | ENST00000384907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002632.hg.1 | NR_030177.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002632.hg.1 | TC16000174.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1600006986.hg.1 | ENST00000384907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006986.hg.1 | NR_030177.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600149530.hg.1 | ENST00000384907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600149530.hg.1 | NR_030177.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600149530.hg.1 | TC1600006986.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600149530.hg.1 | ENST00000384907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600149530.hg.1 | NR_030177.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3308048 | NR_030177.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3308048 | NR_030177.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3308048 | NR_030177.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3308048 | NR_030177.1 | human_transcriptome |

MIR193B
Genomic Map