Assay Details
| Gene Symbol: MIR597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030327.1 | 1 | 39 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC157273 | LOC157273 | -406.6kb | Sense | Chr. 8 - 9325051 - 9335080 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 599 | LINC00599 | Rncr3 | 158.3kb | Anti-sense | Chr. 8 - 9900064 - 9903329 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0800006686.hg.1
This assay targets the same location as HTA transcript cluster(s) TC08000070.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227779 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227783 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227784 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227755 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05948524 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227760 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227747 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227773 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227749 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227751 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05948532 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227778 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227770 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05948556 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227759 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227764 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05948555 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05948543 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227769 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227765 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05948545 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227771 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227757 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05948562 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227768 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227748 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227750 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05948561 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227777 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227762 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05948552 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227773 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05948545 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227747 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227760 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227783 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227764 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05948561 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05948532 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227769 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227771 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05948552 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227765 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227748 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05948556 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05948543 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05948562 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227755 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227779 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227768 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227770 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227749 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227759 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227757 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05948524 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227778 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227762 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227777 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05948555 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227784 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227751 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26227750 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800006686.hg.1 | ENST00000384968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006686.hg.1 | NR_030327.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000668.hg.1 | ENST00000384968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000668.hg.1 | NR_030327.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000668.hg.1 | TC08000070.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0800006686.hg.1 | ENST00000384968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006686.hg.1 | NR_030327.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800145395.hg.1 | ENST00000384968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145395.hg.1 | NR_030327.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800145395.hg.1 | ENST00000384968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145395.hg.1 | NR_030327.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800145395.hg.1 | TC0800006686.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3308642 | NR_030327.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3308642 | NR_030327.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3308642 | NR_030327.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3308642 | NR_030327.1 | human_transcriptome |

MIR597
Genomic Map