Assay Details
| Gene Symbol: MIR4293 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036181.1 | 1 | 44 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FERM domain containing 4A | FRMD4A |
CCAFCA, FRMD4, bA295P9.4 |
-52.3kb | Sense | Chr. 10 - 13643706 - 14330952 on NCBI Build GRCh38 |
| Downstream | microRNA 1265 | MIR1265 |
MIRN1265, hsa-mir-1265 |
53.3kb | Anti-sense | Chr. 10 - 14436576 - 14436661 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1000009859.hg.1
Set Membership: |
|

MIR4293
Genomic Map